2,6-Dimethoxyphenylboronic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

480096-25G 480096-5G
SIAL480096-25GEA 52.1 EUR
SIAL480096-25G SIAL480096-5G
2,6-Dimethoxyphenylboronic acid, Sigma-Aldrich®
2,6-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01318987
CAS Number: 23112-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR