3,4-Dimethoxyphenylboronic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

480118-25G 480118-5G
SIAL480118-25GEA 88.1 EUR
SIAL480118-25G SIAL480118-5G
3,4-Dimethoxyphenylboronic acid, Sigma-Aldrich®
3,4-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01074574
CAS Number: 122775-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR