1,2,4,5-Tetrabromobenzene, Sigma-Aldrich®

Supplier: Merck
Warning

278343-100G 278343-25G
SIAL278343-100GEA 337 EUR
SIAL278343-100G SIAL278343-25G
1,2,4,5-Tetrabromobenzene, Sigma-Aldrich®
1,2,4,5-Tetrabromobenzene
Formula: C₆H₂Br₄
MW: 393.7 g/mol
MDL Number: MFCD00000063
CAS Number: 636-28-2
EINECS: 211-253-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR