1,3,5-Tribromobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
140066-100G 140066-25G
SIAL140066-100GEA 90.1 EUR
SIAL140066-100G SIAL140066-25G
1,3,5-Tribromobenzene, Sigma-Aldrich®
1,3,5-Tribromobenzene
Formula: C₆H₃Br₃
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 122…123 °C
Density: 2.637 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000080
CAS Number: 626-39-1
EINECS: 210-947-3