1,3-Dibromobenzene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
194395-25G 194395-5G
SIAL194395-25GEA 64.6 EUR
SIAL194395-25G SIAL194395-5G
1,3-Dibromobenzene, Sigma-Aldrich®
1,3-Dibromobenzene
Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: –7 °C
Density: 1.952 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000078
CAS Number: 108-36-1
EINECS: 203-574-2