2-Bromobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: o-Bromobenzaldehyde
M-Clarity™ quality level = MQ100
B57001-10G B57001-25G
SIALB57001-10GEA 50.4 EUR
SIALB57001-10G SIALB57001-25G
2-Bromobenzaldehyde, Sigma-Aldrich®
2-Bromobenzaldehyde
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 21 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003300
CAS Number: 6630-33-7
EINECS: 229-622-2