3-Bromobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
B57206-100G B57206-25G
SIALB57206-100GEA 96.2 EUR
SIALB57206-100G SIALB57206-25G
3-Bromobenzaldehyde, Sigma-Aldrich®
Formula: C₇H₅BrO
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: 18…21 °C
Density: 1.587 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00003345
CAS Number: 3132-99-8
EINECS: 221-526-9