4-Bromobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
B57400-100G
B57400-10G
B57400-25G
SIALB57400-100GEA
174
EUR
SIALB57400-100G
SIALB57400-10G
SIALB57400-25G
4-Bromobenzaldehyde, Sigma-Aldrich®
4-Bromobenzaldehyde
Formula:
C₇H₅BrO MW: 185.02 g/mol Boiling Pt: 244 °C (1013 hPa) Melting Pt: 54…56 °C Density: 1.849 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003377 CAS Number: 1122-91-4 EINECS: 214-365-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...