2-Amino-3,5-dibromobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

463469-25G 463469-5G
SIAL463469-25GEA 131 EUR
SIAL463469-25G SIAL463469-5G
2-Amino-3,5-dibromobenzaldehyde, Sigma-Aldrich®
2-Amino-3,5-dibromobenzaldehyde
Formula: C₇H₅Br₂NO
MW: 278.93 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 137…137.5 °C
MDL Number: MFCD00671100
CAS Number: 50910-55-9
EINECS: 256-841-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR