5-Bromosalicylaldehyde ≥98%, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5-Bromo-2-hydroxybenzaldehyde

M-Clarity™ quality level = MQ200

137286-100G 137286-25G
SIAL137286-100GEA 301 EUR
SIAL137286-100G SIAL137286-25G
5-Bromosalicylaldehyde ≥98%, Sigma-Aldrich®
5-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 102…106 °C (lit.)
MDL Number: MFCD00003330
CAS Number: 1761-61-1
EINECS: 217-167-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR