1-Bromo-3,5-dichlorobenzene, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3,5-Dichlorobromobenzene
M-Clarity™ quality level = MQ100
277789-100G
277789-25G
SIAL277789-100GEA
93.1
EUR
SIAL277789-100G
SIAL277789-25G
1-Bromo-3,5-dichlorobenzene, Sigma-Aldrich®
1-Bromo-3,5-dichlorobenzene
Formula:
C₆H₃BrCl₂ MW: 225.9 g/mol Boiling Pt: 227 °C (1013 hPa) Melting Pt: 82…84 °C |
MDL Number:
MFCD00000584 CAS Number: 19752-55-7 EINECS: 243-270-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...