1-Bromo-3-chlorobenzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Bromochlorobenzene
M-Clarity™ quality level = MQ200
124036-100G 124036-25G
SIAL124036-100GEA 153 EUR
SIAL124036-100G SIAL124036-25G
1-Bromo-3-chlorobenzene, Sigma-Aldrich®
1-Bromo-3-chlorobenzene
Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –21.2 °C
Density: 1.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000568
CAS Number: 108-37-2
EINECS: 203-575-8