2-Bromo-5-chlorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
664022-25G
664022-5G
SIAL664022-25GEA
79.2
EUR
SIAL664022-25G
SIAL664022-5G
2-Bromo-5-chlorobenzoic acid, Sigma-Aldrich®
2-Bromo-5-chlorobenzoic acid
Formula:
C₇H₄BrClO₂ MW: 235.46 g/mol |
MDL Number:
MFCD00013982 CAS Number: 21739-93-5 EINECS: 244-559-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...