5-Bromo-2-chlorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
175544-100G 175544-25G
SIAL175544-100GEA 56.6 EUR
SIAL175544-100G SIAL175544-25G
5-Bromo-2-chlorobenzoic acid, Sigma-Aldrich®
5-Bromo-2-chlorobenzoic acid
Formula: C₇H₄BrClO₂
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002415
CAS Number: 21739-92-4
EINECS: 244-558-5