3-Bromobenzhydrazide ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Bromobenzoic hydrazide

A19628.06 A19628.14
A19628.06EA 70.9 EUR
A19628.06 A19628.14
3-Bromobenzhydrazide ≥98%
3-Bromobenzhydrazide

Purity: 98+% CAS No.: 39115-96-3 Molecular Formula: C7H7BrN2O Formula Weight: 215.05 Melting Point: 154-157° Beilstein Reference: 1945814

Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
Boiling Pt: 366
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00007600
CAS Number: 39115-96-3
EINECS: 254-298-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR