Ethyl-3-bromobenzoate 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Ethyl 3-bromobenzoate, 3-Bromobenzoic acid ethyl ester
308540250 308540050
ACRO308540250EA 66.1 EUR
ACRO308540250 ACRO308540050
Ethyl-3-bromobenzoate 98%
Ethyl 3-bromobenzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 130…131 °C (12 mmHg)
Density: 1.43 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013529
CAS Number: 24398-88-7
EINECS: 246-223-9
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5405 to 1.5425 (20°C, 589 nm)
GC ≥97.5 %