2-Bromobenzoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
o-Bromobenzoic Acid
M-Clarity™ quality level = MQ100
137677-100G
137677-25G
SIAL137677-100GEA
66.2
EUR
SIAL137677-100G
SIAL137677-25G
2-Bromobenzoic acid, Sigma-Aldrich®
2-Bromobenzoic acid
Formula:
C₇H₅BrO₂ MW: 201.02 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 144…147 °C Density: 1.929 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002402 CAS Number: 88-65-3 EINECS: 201-848-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...