4-Bromobenzoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
p-Bromobenzoic Acid
M-Clarity™ quality level = MQ100
108510-100G
108510-25G
SIAL108510-100GEA
104
EUR
SIAL108510-100G
SIAL108510-25G
4-Bromobenzoic acid, Sigma-Aldrich®
4-Bromobenzoic acid
Formula:
C₇H₅BrO₂ MW: 201.02 g/mol Boiling Pt: 300 Melting Pt: 252…256 °C Density: 1.894 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002529 CAS Number: 586-76-5 EINECS: 209-581-7 Merck Index: 13,01392 |