5-Bromoanthranilic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Amino-5-bromobenzoic acid
M-Clarity™ quality level = MQ200
260118-25G 260118-5G
SIAL260118-25GEA 52.7 EUR
SIAL260118-25G SIAL260118-5G
5-Bromoanthranilic acid, Sigma-Aldrich®
5-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 342
Melting Pt: 218…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00007823
CAS Number: 5794-88-7
EINECS: 227-338-3
UN: 2811
ADR: 6.1 III
Merck Index: 13,01389