1-Bromo-3,5-difluorobenzene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3,5-Difluorobromobenzene
M-Clarity™ quality level = MQ100
290165-100G 290165-10G
SIAL290165-100GEA 183 EUR
SIAL290165-100G SIAL290165-10G
1-Bromo-3,5-difluorobenzene, Sigma-Aldrich®
1-Bromo-3,5-difluorobenzene
Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 140
Melting Pt: –27 °C
Density: 1.676 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010305
CAS Number: 461-96-1
EINECS: 416-710-2
UN: 1993
ADR: 3 III