1-Bromo-3-fluorobenzene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Bromofluorobenzene
M-Clarity™ quality level = MQ100
B67007-100G B67007-25G B67007-5G
SIALB67007-100GEA 62.2 EUR
SIALB67007-100G SIALB67007-25G SIALB67007-5G
1-Bromo-3-fluorobenzene, Sigma-Aldrich®
1-Bromo-3-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 148
Melting Pt: –8 °C
Density: 1.7081 g/cm³ (20 °C)
Flash Pt: 46
Storage Temperature: Ambient
MDL Number: MFCD00000326
CAS Number: 1073-06-9
EINECS: 214-023-0
UN: 1993
ADR: 3 III