2-Bromo-4-fluorobenzoic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

367931-1G 367931-5G
SIAL367931-1GEA 25 EUR
SIAL367931-1G SIAL367931-5G
2-Bromo-4-fluorobenzoic acid, Sigma-Aldrich®
2-Bromo-4-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 173…175 °C
MDL Number: MFCD00055370
CAS Number: 1006-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR