2-Bromo-5-fluorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
563498-1G
563498-5G
SIAL563498-1GEA
55.6
EUR
SIAL563498-1G
SIAL563498-5G
2-Bromo-5-fluorobenzoic acid, Sigma-Aldrich®
2-Bromo-5-fluorobenzoic acid
Formula:
C₇H₄BrFO₂ MW: 219.01 g/mol |
MDL Number:
MFCD00142874 CAS Number: 394-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...