2-Bromo-4-fluoroanisole, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2-Bromo-4-fluoro-1-methoxybenzene

M-Clarity™ quality level = MQ100

308919-100G 308919-5G
SIAL308919-100GEA 134 EUR
SIAL308919-100G SIAL308919-5G
2-Bromo-4-fluoroanisole, Sigma-Aldrich®
2-Bromo-4-fluoroanisole
Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 274 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00012014
CAS Number: 452-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR