1-Bromo-3-iodobenzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Bromoiodobenzene
M-Clarity™ quality level = MQ200
280097-25G 280097-5G
SIAL280097-25GEA 66.1 EUR
SIAL280097-25G SIAL280097-5G
1-Bromo-3-iodobenzene, Sigma-Aldrich®
1-Bromo-3-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: –9.3 °C
Density: 2.21 g/cm³ (20 °C)
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9