4-Bromo-N,N-dimethylaniline ≥98%
Supplier: Thermo Scientific
Danger
A10854.22 A10854.14
A10854.22EA 86.1 EUR
A10854.22 A10854.14
4-Bromo-N,N-dimethylaniline ≥98%
4-Bromo-N,N-dimethylaniline
Purity: 98+% CAS No.: 586-77-6 Molecular Formula: C8H10BrN Formula Weight: 200.08 Flash Point: 110°(230°F) Melting Point: 52-56° Boiling Point: 263-265° Beilstein Reference: 2206155 MDL No.: MFCD00000093
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 263…265 °C
Melting Pt: 52…56 °C
Density: 1.3218 g/cm³ (99.5 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000093
CAS Number: 586-77-6
EINECS: 209-582-2
UN: 3288
ADR: 6.1,III