4-Bromo-N,N-dimethylaniline, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
242950-100G
242950-25G
SIAL242950-100GEA
101
EUR
SIAL242950-100G
SIAL242950-25G
4-Bromo-N,N-dimethylaniline, Sigma-Aldrich®
4-Bromo-N,N-dimethylaniline
Formula:
C₈H₁₀BrN MW: 200.08 g/mol Boiling Pt: 264 °C (1013 hPa) Melting Pt: 54…55 °C Density: 1.3218 g/cm³ (99.5 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000093 CAS Number: 586-77-6 EINECS: 209-582-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...