3-Bromophenyl isocyanate
Supplier: Thermo Scientific
SB01700.1 SB01700.10 SB01700.50
MAYBSB01700.1EA 0 EUR
MAYBSB01700.1 MAYBSB01700.10 MAYBSB01700.50
3-Bromophenyl isocyanate
3-Bromophenyl isocyanate
Formula: C₇H₄BrNO
Boiling Pt: 220 °C (1013 hPa)
Density: 1.586 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002014
CAS Number: 23138-55-8
UN: 2206
ADR: 6.1,III