2-Bromophenyl isothiocyanate

Supplier: Thermo Scientific

S04160.1 S04160.50
MAYBS04160.1EA 0 EUR
MAYBS04160.1 MAYBS04160.50
2-Bromophenyl isothiocyanate
2-Bromophenyl isothiocyanate
Formula: C₇H₄BrNS
MW: 214.09 g/mol
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.59 g/cm³ (20 °C)
MDL Number: MFCD00004799
CAS Number: 13037-60-0
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR