2,4-Dibromoaniline, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
D38405-100G D38405-25G D38405-5G
SIALD38405-100GEA 229 EUR
SIALD38405-100G SIALD38405-25G SIALD38405-5G
2,4-Dibromoaniline, Sigma-Aldrich®
2,4-Dibromoaniline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 156 °C (21 mmHg)
Melting Pt: 80…82 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007633
CAS Number: 615-57-6
EINECS: 210-434-4
UN: 2811
ADR: 6.1,III