3-Bromoaniline, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

180025-100G 180025-25G
SIAL180025-100GEA 127 EUR
SIAL180025-100G SIAL180025-25G
3-Bromoaniline, Sigma-Aldrich®
3-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1.59 g/cm³ (20 °C)
Flash Pt: >230 °C
Storage Temperature: Ambient
MDL Number: MFCD00007757
CAS Number: 591-19-5
EINECS: 209-704-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR