4-Bromoaniline, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
100900-100G 100900-500G 100900-5G
SIAL100900-100GEA 64.2 EUR
SIAL100900-100G SIAL100900-500G SIAL100900-5G
4-Bromoaniline, Sigma-Aldrich®
4-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 62…64 °C
Density: 1.799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007822
CAS Number: 106-40-1
EINECS: 203-393-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,01388