2,3-Difluoro-6-nitrophenol

Supplier: Thermo Scientific

Synonyms: 1,2-Difluoro-3-hydroxy-4-nitrobenzene

NRB03398.250 NRB03398.1
MAYBNRB03398.250EA 0 EUR
MAYBNRB03398.250 MAYBNRB03398.1
2,3-Difluoro-6-nitrophenol
2,3-Difluoro-6-nitrophenol
Formula: C₆H₃F₂NO₃
MW: 175.09 g/mol
Melting Pt: 61 °C
MDL Number: MFCD00042255
CAS Number: 82419-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR