(2-Bromoethoxy)benzene
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
β-Bromophenetole, 2-Phenoxyethyl bromide, 2-Bromoethyl phenyl ether, ω-Bromophenetole
SB01861.1
SB01861.10
SB01861.50
MAYBSB01861.1EA
0
EUR
MAYBSB01861.1
MAYBSB01861.10
MAYBSB01861.50
(2-Bromoethoxy)benzene
(2-Bromoethoxy)benzene
Formula:
BrCH₂CH₂OC₆H₅ MW: 201.06 g/mol Boiling Pt: 144 °C (53 hPa) Melting Pt: 28…32 °C Density: 1,3555 g/cm³ (20 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000234 CAS Number: 589-10-6 EINECS: 209-634-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...