(2-Bromoethoxy)benzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: β-Bromophenetole, 2-Phenoxyethyl bromide, 2-Bromoethyl phenyl ether, ω-Bromophenetole
M-Clarity™ quality level = MQ100
B75506-100G B75506-25G
SIALB75506-100GEA 288 EUR
SIALB75506-100G SIALB75506-25G
(2-Bromoethoxy)benzene, Sigma-Aldrich®
(2-Bromoethoxy)benzene
Formula: BrCH₂CH₂OC₆H₅
MW: 201.06 g/mol
Boiling Pt: 144 °C (53 hPa)
Melting Pt: 28…32 °C
Density: 1.3555 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000234
CAS Number: 589-10-6
EINECS: 209-634-4