2-Bromoanisole, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Bromo-2-methoxybenzene
M-Clarity™ quality level = MQ200
159239-100G
159239-25G
SIAL159239-100GEA
41.3
EUR
SIAL159239-100G
SIAL159239-25G
2-Bromoanisole, Sigma-Aldrich®
2-Bromoanisole
Formula:
C₇H₇BrO MW: 187.04 g/mol Boiling Pt: 223 °C (1013 hPa) Density: 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000064 CAS Number: 578-57-4 EINECS: 209-425-8 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...