3-Bromoanisole, Sigma-Aldrich®
Supplier: Merck
|
|
Synonyms:
1-Bromo-3-methoxybenzene
M-Clarity™ quality level = MQ200
B56498-100G
B56498-25G
SIALB56498-100GEA
125
EUR
SIALB56498-100G
SIALB56498-25G
3-Bromoanisole, Sigma-Aldrich®
3-Bromoanisole
Formula:
C₇H₇BrO MW: 187.04 g/mol Boiling Pt: 211 °C (1013 hPa) Melting Pt: 2 °C Density: 1.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000081 CAS Number: 2398-37-0 EINECS: 219-264-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...