2-Bromophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
130915-10G
130915-50G
SIAL130915-10GEA
71.7
EUR
SIAL130915-10G
SIAL130915-50G
2-Bromophenol, Sigma-Aldrich®
2-Bromophenol
Formula:
C₆H₅BrO MW: 173.01 g/mol Boiling Pt: 195 °C (1013 hPa) Melting Pt: 5.6 °C Density: 1.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002146 CAS Number: 95-56-7 EINECS: 202-432-7 UN: 3082 ADR: 9,III Merck Index: 13,01413 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...