3-Bromophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
101079-100G 101079-10G 101079-25G
SIAL101079-100GEA 137 EUR
SIAL101079-100G SIAL101079-10G SIAL101079-25G
3-Bromophenol, Sigma-Aldrich®
3-Bromophenol
Formula: C₆H₅BrO
MW: 173.01 g/mol
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 27…30 °C
Density: 0.641 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002253
CAS Number: 591-20-8
EINECS: 209-706-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,01413