4-Bromoresorcinol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Bromobenzene-1,3-diol
M-Clarity™ quality level = MQ100
B80607-25G B80607-5G
SIALB80607-25GEA 498 EUR
SIALB80607-25G SIALB80607-5G
4-Bromoresorcinol, Sigma-Aldrich®
4-Bromoresorcinol
Formula: C₆H₅BrO₂
MW: 189.01 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 98 °C
MDL Number: MFCD00002272
CAS Number: 6626-15-9
EINECS: 229-586-8