4-Bromothioanisole ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: 1-Bromo-4-(methylthio)benzene, 4-Bromophenyl methyl sulphide
A15504.22 A15504.36
A15504.22EA 156 EUR
A15504.22 A15504.36
4-Bromothioanisole ≥98%
4-Bromothioanisole
Purity: 98+% CAS No.: 104-95-0 Molecular Formula: C7H7BrS Formula Weight: 203.11 Flash Point: 110°(230°F) Odour: Stench Boiling Point: 250° Beilstein Reference: 1906693 MDL No.: MFCD00000102
Formula: C₇H₇BrS
MW: 203.1 g/mol
Boiling Pt: 250 °C
Melting Pt: 37…40 °C
Density: 1.4584 g/cm³ (39 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000102
CAS Number: 104-95-0
EINECS: 203-255-8
UN: 3335
ADR: 9,