4-Bromothioanisole, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-Bromo-4-(methylthio)benzene, 4-Bromophenyl methyl sulphide
M-Clarity™ quality level = MQ100
196525-100G
196525-25G
SIAL196525-100GEA
229
EUR
SIAL196525-100G
SIAL196525-25G
4-Bromothioanisole, Sigma-Aldrich®
4-Bromothioanisole
Formula:
C₇H₇BrS MW: 203.1 g/mol Boiling Pt: 242 °C (1013 hPa) Melting Pt: 35 °C Density: 1.4584 g/cm³ (39 °C) |
MDL Number:
MFCD00000102 CAS Number: 104-95-0 EINECS: 203-255-8 UN: 3335 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...