4-Bromothiophenol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 4-Bromobenzenethiol, p-Bromothiophenol, 4-Bromobenzene-1-thiol, 1-Bromo-4-mercaptobenzene

M-Clarity™ quality level = MQ100

B81956-25G B81956-5G
SIALB81956-25GEA 96.6 EUR
SIALB81956-25G SIALB81956-5G
4-Bromothiophenol, Sigma-Aldrich®
4-Bromothiophenol
Formula: C₆H₅BrS
Boiling Pt: 239 °C (1013 hPa)
Melting Pt: 70…71 °C
Density: 1.526 g/cm³ (83 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004845
CAS Number: 106-53-6
EINECS: 203-407-3
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR