2,4-Dibromobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,4-Dibromobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

548677-1G 548677-5G
SIAL548677-1GEA 32.3 EUR
SIAL548677-1G SIAL548677-5G
2,4-Dibromobenzenesulphonyl chloride, Sigma-Aldrich®
2,4-Dibromobenzenesulphonyl chloride
Formula: C₆H₃Br₂ClO₂S
MW: 334.42 g/mol
MDL Number: MFCD03093759
CAS Number: 72256-95-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR