4-Bromobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
108669-100G 108669-25G
SIAL108669-100GEA 101 EUR
SIAL108669-100G SIAL108669-25G
4-Bromobenzenesulphonyl chloride, Sigma-Aldrich®
4-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 74 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007437
CAS Number: 98-58-8
EINECS: 202-683-2
UN: 3261
ADR: 8,II
Merck Index: 13,01391