3-Cyanobenzaldehyde

Supplier: Thermo Scientific
Warning

Synonyms: 3-Formylbenzonitrile

SEW02565DA SEW02565EA
MAYBSEW02565DAEA 0 EUR
MAYBSEW02565DA MAYBSEW02565EA
3-Cyanobenzaldehyde
3-Cyanobenzaldehyde
Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003344
CAS Number: 24964-64-5
EINECS: 246-549-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR