4-Cyanobenzaldehyde
Supplier: Thermo Scientific
Synonyms:
4-Formylbenzonitrile
SB00611.1
SB00611.10
MAYBSB00611.1EA
0
EUR
MAYBSB00611.1
MAYBSB00611.10
4-Cyanobenzaldehyde
4-Cyanobenzaldehyde
Formula:
C₈H₅NO MW: 131.13 g/mol Boiling Pt: 262 °C (1013 hPa) Melting Pt: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003376 CAS Number: 105-07-7 EINECS: 203-267-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...