Vanillic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Hydroxy-3-methoxybenzoic acid
M-Clarity™ quality level = MQ200
H36001-100G H36001-25G
SIALH36001-100GEA 128 EUR
SIALH36001-100G SIALH36001-25G
Vanillic acid, Sigma-Aldrich®
Vanillic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 353
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8
Merck Index: 12,10068