2-Aminobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning

A9628-100MG A9628-500MG A9628-1G
SIALA9628-100MGEA 95 EUR
SIALA9628-100MG SIALA9628-500MG SIALA9628-1G
2-Aminobenzaldehyde, Sigma-Aldrich®
2-Aminobenzaldehyde
Formula: C₇H₇NO
MW: 121.14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00007709
CAS Number: 529-23-7
EINECS: 208-454-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR