4-Nitrobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
MQ100
130176-100G 130176-10G 130176-25G
SIAL130176-100GEA 104 EUR
SIAL130176-100G SIAL130176-10G SIAL130176-25G
4-Nitrobenzaldehyde, Sigma-Aldrich®
4-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 300
Melting Pt: 106
Density: 1.546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007346
CAS Number: 555-16-8
EINECS: 209-084-5
Merck Index: 12,06684